Merck
  • Two new dfr genes in trimethoprim-resistant integron-negative Escherichia coli isolates.
Two new dfr genes in trimethoprim-resistant integron-negative Escherichia coli isolates.
Antimicrobial agents and chemotherapy (2007-02-20)
M Grape, L Sundström, G Kronvall
ABSTRACT
Two Escherichia coli isolates resistant to trimethoprim but negative for integrons carried two new resistance genes, dfrA24 and dfrA26, remotely similar to one another and to the cassette-independent genes dfrA8 and dfrA9. The dfrA24 gene was not associated with known mobile elements, while dfrA26 was associated with the CR1 common region.
Product Description
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Trimethoprim, ≥98.5%