Merck
  • Hollow Zn-Co Based Zeolitic Imidazole Framework as a Robust Heterogeneous Catalyst for Enhanced CO2 Chemical Fixation.
Hollow Zn-Co Based Zeolitic Imidazole Framework as a Robust Heterogeneous Catalyst for Enhanced CO2 Chemical Fixation.
Chemistry, an Asian journal (2019-10-28)
Bo Tang, Shuang Li, Wei-Chao Song, Yan Li, En-Cui Yang, Xiao-Jun Zhao, Landong Li
ABSTRACT
The efficient chemical conversion of carbon dioxide (CO2 ) into value-added fine chemicals is an intriguing but challenging route in sustainable chemistry. Herein, a hollow-structured bimetallic zeolitic imidazole framework composed of Zn and Co as metal centers (H-ZnCo-ZIF) has been successfully prepared via a post-synthetic strategy based on controllable chemical-etching of the preformed solid ZnCo-ZIF in tannic acid. The creation of hollow cavities inside each monocrystalline ZIFs could be achieved without destroying the intrinsic frameworks, as characterized by field-emission scanning electron microscopy, transmission electron microscopy, and X-ray diffraction technologies. The as-synthesized H-ZnCo-ZIF exhibited remarkable catalytic activity in the cycloaddition of CO2 with epoxides to the corresponding cyclic carbonates, outperforming the solid ZnCo-ZIF analogue due to the improved mass transfer originating from the hollow structure. More importantly, due to stabilization of metal centers in the ZIF framework by the tannic acid shell, H-ZnCo-ZIF exhibited good recyclability, and no activity loss could be observed in six runs. The present study provides a simple and effective strategy to enhance the catalytic performance of ZIFs by creating a hollow structure via chemical etching.
Sigma-Aldrich
1,2-Epoxy-3-phenoxypropane, 99%
Sigma-Aldrich
Cyclooctene oxide, 99%
Sigma-Aldrich
Tetraethylammonium bromide, ReagentPlus®, 99%
Sigma-Aldrich
Cyclohexene oxide, 98%