Merck
  • C6 Ceramide (d18:1/6:0) as a Novel Treatment of Cutaneous T Cell Lymphoma.
C6 Ceramide (d18:1/6:0) as a Novel Treatment of Cutaneous T Cell Lymphoma.
Cancers (2021-01-17)
Raphael Wilhelm, Timon Eckes, Gergely Imre, Stefan Kippenberger, Markus Meissner, Dominique Thomas, Sandra Trautmann, Jean-Philippe Merlio, Edith Chevret, Roland Kaufmann, Josef Pfeilschifter, Alexander Koch, Manuel Jäger
ABSTRACT
Cutaneous T cell lymphomas (CTCLs) represent a heterogeneous group of T cell lymphomas that primarily affect the skin. The most frequent forms of CTCL are mycosis fungoides and Sézary syndrome. Both are characterized by frequent recurrence, developing chronic conditions and high mortality with a lack of a curative treatment. In this study, we evaluated the effect of short-chain, cell-permeable C6 Ceramide (C6Cer) on CTCL cell lines and keratinocytes. C6Cer significantly reduced cell viability of CTCL cell lines and induced cell death via apoptosis and necrosis. In contrast, primary human keratinocytes and HaCaT keratinocytes were less affected by C6Cer. Both keratinocyte cell lines showed higher expressions of ceramide catabolizing enzymes and HaCaT keratinocytes were able to metabolize C6Cer faster and more efficiently than CTCL cell lines, which might explain the observed protective effects. Along with other existing skin-directed therapies, C6Cer could be a novel well-tolerated drug for the topical treatment of CTCL.
Sigma-Aldrich
Carmofur, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-ASAH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution