Merck
  • Knockdown of CRAD suppresses the growth and promotes the apoptosis of human lung cancer cells via Claudin 4.
Knockdown of CRAD suppresses the growth and promotes the apoptosis of human lung cancer cells via Claudin 4.
Bioscience reports (2020-10-03)
Anfang Cui, Yuchan Xue, Xi'ao Wang, Yanhong Huang, Xiaolin Han, Xiangling Li, Delei Niu, Shaorui Niu, Yujie Zhao, Xinyu Yang, Wei Yu
ABSTRACT
Non-small cell lung cancer (NSCLC) is one of the most common causes of cancer-related mortality globally. However, the mechanism underlying NSCLC is not fully understood. Here, we investigated the role of cancer-related regulator of actin dynamics (CRAD) in NSCLC. We showed that CRAD was up-regulated in human NSCLC tissues and lung cancer cell lines. Lentivirus-mediated knockdown of CRAD repressed the proliferation and colony growth of A549 and H1299 cells. Apoptosis was enhanced by CRAD silencing in both cells, implicating that CRAD might maintain the survival of lung cancer cells. Microarray and bioinformatic assay revealed that CRAD directly or indirectly regulated diverse genes, including those involved in cell cycle and DNA damage repair. qRT-PCR and Western blot results confirmed the dysregulated genes as shown in microarray analysis. Claudin 4 was up-regulated in CRAD silenced A549 cells. The knockdown of Claudin 4 blocked the effects of CRAD on the expression of cell cycle and apoptosis effectors and enhanced the viability of A549 cells with CRAD down-regulation. Taken together, our findings demonstrate that CRAD acts as an oncogene in NSCLC at least partly through repressing Claudin 4.
Sigma-Aldrich
Anti-KIAA1211 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution