Merck
  • Lidocaine alleviates cisplatin resistance and inhibits migration of MGC-803/DDP cells through decreasing miR-10b.
Lidocaine alleviates cisplatin resistance and inhibits migration of MGC-803/DDP cells through decreasing miR-10b.
Cell cycle (Georgetown, Tex.) (2020-09-08)
Xiaomin Zhang, Guangfeng Gu, Xuanfei Li, Chaopei Zhang
ABSTRACT
Although chemotherapy is one of the effective means of treating gastric cancer, the resistance of chemotherapeutic drugs has followed. And the mechanisms of resistance are not completely clear. The main aim of this article was to develop a kind of drug that could reduce the resistance of cisplatin on gastric cancer cells. The MGC-803 and MGC-803/DDP cells were treated by cisplatin for 48 h and Lidocaine (Lido) for 24 h. Cell viability, apoptosis, migration and invasion were tested by cell counting kit-8 (CCK-8) assay, apoptosis assay, western blot, migration and invasion assay. After MGC-803/DDP cells were transfected for 48 h, the expression of microRNA-10b (miR-10b) were detected by quantitative reverse transcriptase-polymerase chain reaction (qRT-PCR). Activation of AKT/mTOR and β-catenin pathways was tested by western blot. Cisplatin caused MGC-803 and MGC-803/DDP cell apoptosis, and MGC-803/DDP cells possessed higher cisplatin resistance than MGC-803 cells. Lido reduced the cisplatin resistance of MGC-803/DDP cells. Besides, Lido inhibited MGC-803/DDP cell migration and invasion. In addition, Lido declined cisplatin resistance by down-regulating miR-10b. Lido also repressed AKT/mTOR and β-catenin pathway by down-regulating miR-10b. This article explained the role of Lido in cisplatin resistance in MGC-803/DDP cells. Furthermore, Lido weakened the cisplatin resistance in MGC-803/DDP cells at least in part through decreasing the expression of miR-10b.
Sigma-Aldrich
Monoclonal Anti-TIMP1 antibody produced in mouse, clone 4D12, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder