Merck
  • Cyrene™ as an Alternative Sustainable Solvent for the Preparation of Poly(lactic-co-glycolic acid) Nanoparticles.
Cyrene™ as an Alternative Sustainable Solvent for the Preparation of Poly(lactic-co-glycolic acid) Nanoparticles.
Journal of pharmaceutical sciences (2020-12-05)
Christian Grune, Jana Thamm, Oliver Werz, Dagmar Fischer
ABSTRACT
Toxic and environmental harmful organic solvents are widely applied to prepare poly(lactic-co-glycolic acid) (PLGA)-based nanoparticles (NP) in standard preparation methods. Alternative non-toxic solvents suffer from disadvantages like high viscosity and plasticizing effects. To overcome these hurdles, Cyrene™ as a new sustainable, non-toxic and low viscous solvent was used to formulate PLGA NPs. A new preparation method was developed and optimized. Small sized blank NPs around 220 nm with a narrow size distribution and highly negative charge (<-23 mV) were obtained. To test the application for drug delivery, the lipophilic model drug atorvastatin was encapsulated in high drug loads with comparable physicochemical characteristics as the blank NPs, and a total drug release within 24 h. No changes of the crystallinity or plasticizing effects could be observed. Highly purified NPs were obtained with a residual Cyrene™ content <2.5%. Finally, the biocompatibility of Cyrene™ itself and of the NPs formed in the presence of Cyrene™ was demonstrated in a hen's egg test. Conclusively, the use of Cyrene™ as solvent offers a simple, fast and non-toxic procedure for preparation of PLGA NPs as drug delivery systems circumventing the downsides of standard methods.
Sigma-Aldrich
Cyrene, BioRenewable, DMF and NMP Substitute