Merck
Have the bloody cells gone to our heads?
The Journal of cell biology (2001-11-29)
K Meletis, J Frisen
ABSTRACT
Recent studies have shown that cells expressing neuronal antigens can be derived from a bone marrow transplant. A new report lends support to and extends these previous results by presenting compelling morphological evidence for the generation and integration of highly differentiated bone marrow-derived neurons.