Merck
  • miR-370 Sensitizes TMZ Response Dependent of MGMT Status in Primary Central Nervous System Lymphoma.
miR-370 Sensitizes TMZ Response Dependent of MGMT Status in Primary Central Nervous System Lymphoma.
Pathology oncology research : POR (2019-02-04)
Xinwei Li, Xueying Xu, Keng Chen, Haijian Wu, Yirong Wang, Shuxu Yang, Kun Wang
ABSTRACT
Primary central nervous system lymphoma (PCNSL) is an aggressive and rare subtype of non-Hodgkin lymphoma, arising exclusively in the CNS with a poor prognosis. Previous evidence has proved that MGMT was a promising target involving in TMZ resistance of PCNSL. Our study described a new miR-370-mediated mechanism of MGMT regulation in PCNSL. We first showed that miR-370 was downregulated in PCNSL tissues, while MGMT was inversely overexpressed. It was also observed that miR-370 suppressed the expression of MGMT. Additionally, upregulation of miR-370 significantly increased TMZ sensitivity dependent of MGMT, thus suppressed Raji cell proliferation and induced apoptosis in vitro. In conclusion, these results suggest that miR-370 is a potential target in PCNSL treatment.
Sigma-Aldrich
MISSION® esiRNA, targeting human MGMT