Merck
  • Agmatine reverses memory deficits induced by Aβ1-42 peptide in mice: A key role of imidazoline receptors.
Agmatine reverses memory deficits induced by Aβ1-42 peptide in mice: A key role of imidazoline receptors.
Pharmacology, biochemistry, and behavior (2020-07-01)
Nandkishor Kotagale, Madhura Dixit, Harshal Garmelwar, Shraddha Bhondekar, Milind Umekar, Brijesh Taksande
ABSTRACT
Agmatine is a biogenic amine synthesized following decarboxylation of L-arginine by the enzyme arginine decarboxylase and exhibits favourable outcome in neurodegenerative disorders. Present study was designed to examine the relationship between agmatine and the imidazoline receptors in memory deficits induced by Aβ1-42 peptide in mice. Mice were treated with single intracerebroventricular (i.c.v.) injection of Aβ1-42 peptide (3 μg) and evaluated for learning and memory in Morris water maze (MWM) and subjected to Aβ1-42, TNF-α and IL-6 and BDNF immunocontent analysis within the hippocampus. While the learning and memory impairment was evident in the mice subjected to MWM test following Aβ1-42 peptide administration, there was a significant increase in Aβ1-42, TNF-α and IL-6 and reduction in BDNF immunocontent within the hippocampus. Daily intraperitoneal (i.p.) treatment with agmatine (10 and 20 mg/kg); imidazoline I1 receptor agonist, moxonidine and imidazoline I2 receptor agonist, 2-BFI starting from day 8 to 27 post-Aβ1-42 injection, significantly prevented the cognitive deficits and normalized the Aβ1-42 peptide, IL-6, TNF-α and BDNF immunocontent in hippocampus. On the other hand, pre-treatment with imidazoline I1 receptor antagonist, efaroxan and imidazoline I2 receptor antagonist, BU 224 attenuated the effects of agmatine on learning and memory in MWM, IL-6, TNF-α and BDNF content. In conclusion, the present study provides functional evidence for the involvement of the imidazoline receptors in agmatine induced reversal of Aβ1-42 induced memory deficits in mice. The data projects agmatine and imidazoline receptor agonists as a potential therapeutic target for the treatment of AD.
Sigma-Aldrich
Moxonidine hydrochloride, ≥98%
Sigma-Aldrich
Efaroxan hydrochloride, ≥98% (HPLC)