Merck
  • Discovery of a tetracyclic quinoxaline derivative as a potent and orally active multifunctional drug candidate for the treatment of neuropsychiatric and neurological disorders.
Discovery of a tetracyclic quinoxaline derivative as a potent and orally active multifunctional drug candidate for the treatment of neuropsychiatric and neurological disorders.
Journal of medicinal chemistry (2014-02-25)
Peng Li, Qiang Zhang, Albert J Robichaud, Taekyu Lee, John Tomesch, Wei Yao, J David Beard, Gretchen L Snyder, Hongwen Zhu, Youyi Peng, Joseph P Hendrick, Kimberly E Vanover, Robert E Davis, Sharon Mates, Lawrence P Wennogle
ABSTRACT
We report the synthesis and structure-activity relationships of a class of tetracyclic butyrophenones that exhibit potent binding affinities to serotonin 5-HT(2A) and dopamine D2 receptors. This work has led to the discovery of 4-((6bR,10aS)-3-methyl-2,3,6b,9,10,10a-hexahydro-1H,7H-pyrido[3',4':4,5]pyrrolo[1,2,3-de]quinoxalin-8-yl)-1-(4-fluorophenyl)-butan-1-one 4-methylbenzenesulfonate (ITI-007), which is a potent 5-HT(2A) antagonist, postsynaptic D2 antagonist, and inhibitor of serotonin transporter. This multifunctional drug candidate is orally bioavailable and exhibits good antipsychotic efficacy in vivo. Currently, this investigational new drug is under clinical development for the treatment of neuropsychiatric and neurological disorders.
Sigma-Aldrich
Lumateperone tosylate, ≥98% (HPLC)
Pricing and availability is not currently available.