- Reorientation of cellulose nanowhiskers in agarose hydrogels under tensile loading.
Reorientation of cellulose nanowhiskers in agarose hydrogels under tensile loading.
Agarose hydrogels filled with cellulose nanowhiskers were strained in uniaxial stretching under different humidity conditions. The orientation of the cellulose whiskers was examined before and after testing with an X-ray laboratory source and monitored in situ during loading by synchrotron X-ray diffraction. The aim of this approach was to determine the process parameters for reorienting the cellulose nanowhiskers toward a preferential direction. Results show that a controlled drying of the hydrogel is essential to establish interactions between the matrix and the cellulose nanowhiskers which allow for a stress transfer during stretching and thereby promote their alignment. Rewetting of the sample after reorientation of the cellulose nanowhiskers circumvents a critical increase of stress. This improves the extensibility of the hydrogel and is accompanied by a further moderate alignment of the cellulose nanowhiskers. Following this protocol, cellulose nanowhiskers with an initial random distribution can be reoriented toward a preferential direction, creating anisotropic nanocomposites.