Merck
  • Tanycyte-Like Cells Derived From Mouse Embryonic Stem Culture Show Hypothalamic Neural Stem/Progenitor Cell Functions.
Tanycyte-Like Cells Derived From Mouse Embryonic Stem Culture Show Hypothalamic Neural Stem/Progenitor Cell Functions.
Endocrinology (2019-05-29)
Mayuko Kano, Hidetaka Suga, Takeshi Ishihara, Mayu Sakakibara, Mika Soen, Tomiko Yamada, Hajime Ozaki, Kazuki Mitsumoto, Takatoshi Kasai, Mariko Sugiyama, Takeshi Onoue, Taku Tsunekawa, Hiroshi Takagi, Daisuke Hagiwara, Yoshihiro Ito, Shintaro Iwama, Motomitsu Goto, Ryoichi Banno, Hiroshi Arima
ABSTRACT
Tanycytes have recently been accepted as neural stem/progenitor cells in the postnatal hypothalamus. Persistent retina and anterior neural fold homeobox (Rax) expression is characteristic of tanycytes in contrast to its transient expression of whole hypothalamic precursors. In this study, we found that Rax+ residual cells in the maturation phase of hypothalamic differentiation in mouse embryonic stem cell (mESC) cultures had similar characteristics to ventral tanycytes. They expressed typical neural stem/progenitor cell markers, including Sox2, vimentin, and nestin, and differentiated into mature neurons and glial cells. Quantitative RT-PCR analysis showed that Rax+ residual cells expressed Fgf-10, Fgf-18, and Lhx2, which are expressed by ventral tanycytes. They highly expressed tanycyte-specific genes Dio2 and Gpr50 compared with Rax+ early hypothalamic progenitor cells. Therefore, Rax+ residual cells in the maturation phase of hypothalamic differentiation were considered to be more differentiated and similar to late progenitor cells and tanycytes. They self-renewed and formed neurospheres when cultured with exogenous FGF-2. Additionally, these Rax+ neurospheres differentiated into three neuronal lineages (neurons, astrocytes, and oligodendrocytes), including neuropeptide Y+ neuron, that are reported to be differentiated from ventral tanycytes toward the arcuate nuclei. Thus, Rax+ residual cells were multipotent neural stem/progenitor cells. Rax+ neurospheres were stably passaged and retained high Sox2 expression even after multiple passages. These results suggest the successful induction of Rax+ tanycyte-like cells from mESCs [induced tanycyte-like (iTan) cells]. These hypothalamic neural stem/progenitor cells may have potential in regenerative medicine and as a research tool.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Vimentin Antibody, serum, Chemicon®