Merck
  • Human Virus-Derived Small RNAs Can Confer Antiviral Immunity in Mammals.
Human Virus-Derived Small RNAs Can Confer Antiviral Immunity in Mammals.
Immunity (2017-06-22)
Yang Qiu, Yanpeng Xu, Yao Zhang, Hui Zhou, Yong-Qiang Deng, Xiao-Feng Li, Meng Miao, Qiang Zhang, Bo Zhong, Yuanyang Hu, Fu-Chun Zhang, Ligang Wu, Cheng-Feng Qin, Xi Zhou
ABSTRACT
RNA interference (RNAi) functions as a potent antiviral immunity in plants and invertebrates; however, whether RNAi plays antiviral roles in mammals remains unclear. Here, using human enterovirus 71 (HEV71) as a model, we showed HEV71 3A protein as an authentic viral suppressor of RNAi during viral infection. When the 3A-mediated RNAi suppression was impaired, the mutant HEV71 readily triggered the production of abundant HEV71-derived small RNAs with canonical siRNA properties in cells and mice. These virus-derived siRNAs were produced from viral dsRNA replicative intermediates in a Dicer-dependent manner and loaded into AGO, and they were fully active in degrading cognate viral RNAs. Recombinant HEV71 deficient in 3A-mediated RNAi suppression was significantly restricted in human somatic cells and mice, whereas Dicer deficiency rescued HEV71 infection independently of type I interferon response. Thus, RNAi can function as an antiviral immunity, which is induced and suppressed by a human virus, in mammals.
Product Description
Roche
DIG Luminescent Detection Kit, sufficient for 50 blots (10 cm x 10 cm each), kit of 1 (5 components), suitable for hybridization
Sigma-Aldrich
Anti-pan Ago Antibody, clone 2A8, clone 2A8, from mouse