Merck
  • Ras-MEK Signaling Mediates a Critical Chk1-Dependent DNA Damage Response in Cancer Cells.
Ras-MEK Signaling Mediates a Critical Chk1-Dependent DNA Damage Response in Cancer Cells.
Molecular cancer therapeutics (2017-02-01)
Ho-June Lee, Yi Cao, Victoria Pham, Elizabeth Blackwood, Catherine Wilson, Marie Evangelista, Christiaan Klijn, David Stokoe, Jeff Settleman
ABSTRACT
Cancer cell line profiling to identify previously unrecognized kinase dependencies revealed a novel nonmutational dependency on the DNA damage response checkpoint kinase Chk1. Although Chk1 is a promising therapeutic target in p53-deficient cancers, we found that Ras-MEK signaling engages Chk1 in a subset of osteosarcoma, ovarian, and breast cancer cells to enable their survival upon DNA damage, irrespective of p53 mutation status. Mechanistically, Ras-MEK signaling drives Chk1 expression and promotes cancer cell growth that produces genotoxic stress that requires Chk1 to mediate a response to the consequent DNA damage. Reciprocally, Chk1 engages a negative feedback loop to prevent hyperactivation of Ras-MEK signaling, thereby limiting DNA damage. Furthermore, exogenous DNA damage promotes Chk1 dependency, and pharmacologic Chk1 inhibition combined with genotoxic chemotherapy potentiates a DNA damage response and tumor cell killing. These findings reveal a mechanism-based diagnostic strategy to identify cancer patients that may benefit from Chk1-targeted therapy. Mol Cancer Ther; 16(4); 694-704. ©2017 AACR.
Sigma-Aldrich
Anti-H-Ras Antibody, clone 7D7.2, clone 7D7.2, Chemicon®, from mouse