Merck
  • Epidemiological, clinical and genetic characterization of aplastic anemia patients in Pakistan.
Epidemiological, clinical and genetic characterization of aplastic anemia patients in Pakistan.
Annals of hematology (2018-11-15)
Zaineb Akram, Parvez Ahmed, Sachiko Kajigaya, Tariq Mahmood Satti, Humayoon Shafique Satti, Qamar Un Nisa Chaudhary, Fernanda Gutierrez-Rodrigues, Pilar F Ibanez, Xingmin Feng, Syed Kamran Mahmood, Tariq Ghafoor, Nighat Shahbaz, Mehreen Ali Khan, Aneesa Sultan
ABSTRACT
Aplastic anemia (AA) is the most serious non-malignant blood disorder in Pakistan, ranked second in prevalence, after thalassemia. We investigated various epidemiological, clinical, and genetic factors of AA in a Pakistani cohort of 214 patients reporting at our hospital between June 2014 and December 2015. A control group of 214 healthy subjects was included for comparison of epidemiological and clinical features. Epidemiological data revealed 2.75-fold higher frequency of AA among males. A single peak of disease onset was observed between ages 10 and 29 years followed by a steady decline. AA was strongly associated with lower socioeconomic profile, rural residence, and high rate of consanguineous marriages. Serum granulocyte colony-stimulating factor and thrombopoietin levels were significantly elevated in AA patients, compared to healthy controls (P < 0.0001), while there was no statistical significance in other nine cytokine levels screened. Allele frequencies of DRB1*15 (56.8%) and DQB1*06 (70.3%) were predominantly high in AA patients. Ten mutations were found in TERT and TERC genes, including two novel mutations (Val526Ala and Val777Met) in exons 3 and 7 of TERT gene. Despite specific features of the AA cohort, this study suggests that epidemiologic and etiologic factors as well as host genetic predisposition exclusively or cooperatively trigger AA in Pakistan.