Merck
Melanogenesis in uveal melanoma cells: Effect of argan oil.
International journal of molecular medicine (2017-08-30)
Nunzia Caporarello, Melania Olivieri, Martina Cristaldi, Dario Rusciano, Gabriella Lupo, Carmelina Daniela Anfuso
ABSTRACT
The mechanisms underlying cutaneous melanogenesis have been widely studied; however, very little is known about uveal melanogenesis. Melanin is normally produced by uveal melanocytes and gives the color to the iris. A derangement from this normal production may occur, for instance, by iatrogenic events, such as glaucoma therapy with prostaglandins that may enhance cutaneous and iris pigmentation. In this study, we investigated the mechanisms that regulate uveal melanogenesis in human uveal melanoma cells (92.1) and murine cutaneous melanoma cells (B16-F1). In the first part of the study, we compared the effects of known cutaneous pigmenting agents on the B16-F1 and 92.1 cells, showing an opposite response of the two cell lines. Subsequently, using argan oil, a known depigmenting agent for murine cutaneous melanoma cells, on 92.1 cells, we found that in these cells, it also functioned as an inhibitor of melanogenesis and tyrosinase expression. From a molecular perspective, treatment of the 92.1 cells with argan oil decreased melanogenesis-associated transcription factor (MITF) gene expression by inducing MITF phosphorylation at Ser73, thus leading to MITF ubiquitination and disposal. It also led to the downregulation of the extracellular signal-regulated kinase (ERK)1/2 and Akt pathways, also known to be involved in cutaneous melanogenesis, although with an opposing function. Taken together, our data indicate that: ⅰ) some differences exist in the regulation of melanogenesis between cutaneous and uveal melanoma cells; and ⅱ) argan oil exerts a depigmenting effect on 92.1 cells through its action on the ERK1/2 and Akt pathways.
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
PD 98,059, solid
Sigma-Aldrich
Anti-phospho-MITF (pSer180/73) antibody produced in rabbit, affinity isolated antibody