Merck
  • Cholinesterase activity in brain of senescence-accelerated-resistant mouse SAMR1 and its variation in brain of senescence-accelerated-prone mouse SAMP8.
Cholinesterase activity in brain of senescence-accelerated-resistant mouse SAMR1 and its variation in brain of senescence-accelerated-prone mouse SAMP8.
Journal of neuroscience research (2009-07-18)
Francisco José Fernández-Gómez, Encarnación Muñoz-Delgado, María Fernanda Montenegro, Francisco J Campoy, Cecilio J Vidal, Joaquín Jordán
ABSTRACT
The early-onset, irreversible, severe deficits of learning and memory in the senescence-accelerated mouse (SAM)-prone/8 (SAMP8) support its use as an animal model for human dementias of early onset. Possible implication of acetylcholinesterase (AChE) and butyrylcholinesterase (BuChE) in cognitive dysfunction of SAMP8 mice was studied by comparing cholinesterase (ChE) expression in brains of SAMP8 mice and of their normal control, SAM-resistant/1 (SAMR1) mice. The level of ChE mRNAs was the same in SAMP8 and SAMR1 brains, which agreed with their equal AChE activity (3.09 +/- 1.45 vs. 3.07 +/- 1.44 mumol.hr(-1).mg protein(-1), U/mg), but not with a doubled BuChE activity in SAMP8 brain (0.14 +/- 0.05 vs. 0.07 +/- 0.02 U/mg; P < 0.01). This great increase in neural BuChE activity may contribute to cognitive deficit of SAMP8 mice. Hydrophilic (G(4) (H), 8%) and amphiphilic (G(4) (A), 74%) AChE tetramers, besides dimers and monomers (G(2) (A) + G(1) (A), 18%), were identified in SAMR1 brains. They also contained G(4) (H) BuChE forms (18%) as well as G(4) (A) (53%) and G(2) (A) + G(1) (A) (29%) species. Although SAMP8 brain displayed proportions of AChE and BuChE forms that were similar to those of SAMR1 brain, phenyl-agarose chromatography with detergent-free extracts showed a rise in the proportion of secretory G(4) (H) BuChE from 35% in SAMR1 to 44% in SAMP8 brain. The strong immunolabelling of glial fibrillary acidic protein (GFAP), a marker of reactive gliosis, in SAMP8 brain and the consideration of BuChE as a marker of glial cells suggest a relationship between phenotypic changes in neuroglial cells and the excess of BuChE activity in SAMP8 brain.
Sigma-Aldrich
Phenyl-Agarose, saline suspension