Merck
  • PP2A activation alone and in combination with cisplatin decreases cell growth and tumor formation in human HuH6 hepatoblastoma cells.
PP2A activation alone and in combination with cisplatin decreases cell growth and tumor formation in human HuH6 hepatoblastoma cells.
PloS one (2019-04-11)
Laura L Stafman, Adele P Williams, Raoud Marayati, Jamie M Aye, Jerry E Stewart, Elizabeth Mroczek-Musulman, Elizabeth A Beierle
ABSTRACT
Despite an increase in incidence, treatments for hepatoblastoma remain virtually unchanged for the past 20 years, emphasizing the need for novel therapeutics. FTY720 (fingolimod) is an immunomodulator approved for use in multiple sclerosis in children that has been demonstrated to have anti-cancer properties in multiple cancer types. We have demonstrated that FTY720 activates PP2A in hepatoblastoma, but does not do so via inhibition of the endogenous inhibitors, CIP2A and I2PP2A, as previously observed in other cancers. PP2A activation in hepatoblastoma decreased cell viability, proliferation, and motility and induced apoptosis. In a subcutaneous xenograft model, FTY720 decreased tumor growth. FTY720 in combination with the standard chemotherapeutic, cisplatin, decreased proliferation in a synergistic manner. Finally, animals bearing subcutaneous hepatoblastoma xenografts treated with FTY720 and cisplatin in combination had significantly decreased tumor growth compared to those treated with either drug alone. These findings show that targeting PP2A with FTY70 shows promise in the treatment of hepatoblastoma and that combining FTY720 with cisplatin may be a novel and effective strategy to better treat this devastating pediatric liver tumor.
Sigma-Aldrich
Anti-phospho-Erk1/2 (Thr202/Tyr204, Thr185/Tyr187)Antibody, recombinant clone AW39R, rabbit monoclonal, clone AW39R, Upstate®, from rabbit
Sigma-Aldrich
Anti-PARP (214/215) cleavage site Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture