Merck
  • A mouse model of subacute liver failure with ascites induced by step-wise increased doses of (-)-epigallocatechin-3-gallate.
A mouse model of subacute liver failure with ascites induced by step-wise increased doses of (-)-epigallocatechin-3-gallate.
Scientific reports (2019-12-04)
Xiaoxiao Wang, Lumin Yang, Jiajia Wang, Yafei Zhang, Ruixia Dong, Ximing Wu, Chung S Yang, Zhenhua Zhang, Jinsong Zhang
ABSTRACT
Acute liver failure is divided into hyperacute, acute and subacute liver failure. Ascites is a common complication of subacute liver failure. Although animal models of acute liver failure have been established, the study of the pathogenesis of subacute liver failure with ascites complication is hampered by the lack of experimental animal model. The present study aimed at providing a mouse model of subacute liver failure with ascites complication. Kunming mice were intraperitoneally injected with (-)-epigallocatechin-3-gallate (EGCG), a redox-active polyphenol from green tea, for 32 consecutive days with step-wise increased dosage. The EGCG treatment resulted in liver failure as evidenced by extensive hepatocyte necrosis observed histologically along with significant elevation of serum alanine aminotransferase, aspartate aminotransferase, total bilirubin and direct bilirubin levels as well as significant reduction of serum albumin. Liver fibrosis was not observed by Masson staining and fibrosis-associated proteins were not increased. The mortality was less than 12% and the survival mice developed noticeable ascites. Hepatic thioredoxin and glutathione systems were activated by the EGCG. These adaptive responses might render most mice tolerable to the EGCG treatment. The EGCG treatment significantly up-regulated renal urea transporter A1 and promoted its trafficking to apical membrane. These alterations, known to increase water reabsorption, may be responsible, at least in part, for the formation of the ascites. Overall, the mice treated with gradually elevated doses of EGCG exhibits some of the features observed in patients with subacute liver failure, especially ascites. This mouse model is a useful tool for investigating the pathogenesis of subacute liver failure with ascites complication.
Sigma-Aldrich
Anti-SLC14A2 antibody produced in rabbit, 1 mg/mL, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid