Merck
  • Functional Analysis of the Replication Fork Proteome Identifies BET Proteins as PCNA Regulators.
Functional Analysis of the Replication Fork Proteome Identifies BET Proteins as PCNA Regulators.
Cell reports (2019-09-26)
Sarah R Wessel, Kareem N Mohni, Jessica W Luzwick, Huzefa Dungrawala, David Cortez
ABSTRACT
Identifying proteins that function at replication forks is essential to understanding DNA replication, chromatin assembly, and replication-coupled DNA repair mechanisms. Combining quantitative mass spectrometry in multiple cell types with stringent statistical cutoffs, we generated a high-confidence catalog of 593 proteins that are enriched at replication forks and nascent chromatin. Loss-of-function genetic analyses indicate that 85% yield phenotypes that are consistent with activities in DNA and chromatin replication or already have described functions in these processes. We illustrate the value of this resource by identifying activities of the BET family proteins BRD2, BRD3, and BRD4 in controlling DNA replication. These proteins use their extra-terminal domains to bind and inhibit the ATAD5 complex and thereby control the amount of PCNA on chromatin.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
5-Iodo-2′-deoxyuridine, ≥99% (HPLC)
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human BRD3
Sigma-Aldrich
MISSION® esiRNA, targeting human ATAD5
Sigma-Aldrich
MISSION® esiRNA, targeting human BRD4
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse