Merck
  • Identification of Atuveciclib (BAY 1143572), the First Highly Selective, Clinical PTEFb/CDK9 Inhibitor for the Treatment of Cancer.
Identification of Atuveciclib (BAY 1143572), the First Highly Selective, Clinical PTEFb/CDK9 Inhibitor for the Treatment of Cancer.
ChemMedChem (2017-09-30)
Ulrich Lücking, Arne Scholz, Philip Lienau, Gerhard Siemeister, Dirk Kosemund, Rolf Bohlmann, Hans Briem, Ildiko Terebesi, Kirstin Meyer, Katja Prelle, Karsten Denner, Ulf Bömer, Martina Schäfer, Knut Eis, Ray Valencia, Stuart Ince, Franz von Nussbaum, Dominik Mumberg, Karl Ziegelbauer, Bert Klebl, Axel Choidas, Peter Nussbaumer, Matthias Baumann, Carsten Schultz-Fademrecht, Gerd Rühter, Jan Eickhoff, Michael Brands
ABSTRACT
Selective inhibition of exclusively transcription-regulating PTEFb/CDK9 is a promising new approach in cancer therapy. Starting from lead compound BAY-958, lead optimization efforts strictly focusing on kinase selectivity, physicochemical and DMPK properties finally led to the identification of the orally available clinical candidate atuveciclib (BAY 1143572). Structurally characterized by an unusual benzyl sulfoximine group, BAY 1143572 exhibited the best overall profile in vitro and in vivo, including high efficacy and good tolerability in xenograft models in mice and rats. BAY 1143572 is the first potent and highly selective PTEFb/CDK9 inhibitor to enter clinical trials for the treatment of cancer.
Sigma-Aldrich
BAY-958 hydrochloride, ≥98% (HPLC)