- A Kinetic Approach to Determining Drug Distribution in Complex Biphasic Systems.
A Kinetic Approach to Determining Drug Distribution in Complex Biphasic Systems.
Pharmaceutical emulsions contain multiple components, such as micellar, aqueous, and oil phases, leading to complex drug transfer and equilibrium phenomena. These complex components present challenges for the bioequivalence assessment of the drug products. The objective of the study was to develop a method that can probe the underlying mechanism and process of drug distribution. The concept of drug partitioning into biphasic systems was used to simplify the complex transfer phenomenon. A kinetic method was developed taking into account the biphasic diffusion. Using this approach, both the rate (kinetics) and the extent (equilibrium) of distribution can be determined. For method development purpose, 3 model compounds (triamcinolone acetonide, difluprednate, and cyclosporine), with expected partition coefficient values ranging from 2 to 6, were tested using the kinetic method and the traditional shake-flask method. The values obtained by the 2 methods for all compounds correlated well (r2 = 0.825). Various organic and aqueous solvents which are commonly encountered in formulations were also tested to determine the impact of phase composition on drug distribution. The kinetic method was found to offer more flexibility in terms of solvent composition and can lead to better understanding for drug distribution and potential drug release in complex biphasic systems.