Merck
  • Can Tenax Extraction Be Used as a Surrogate Exposure Metric for Laboratory-Based Bioaccumulation Tests Using Marine Sediments?
Can Tenax Extraction Be Used as a Surrogate Exposure Metric for Laboratory-Based Bioaccumulation Tests Using Marine Sediments?
Environmental toxicology and chemistry (2019-02-23)
Federico L Sinche, Guilherme R Lotufo, Peter Landrum, Michael J Lydy
ABSTRACT
The Tenax technique was used as an alternative exposure metric to assess the bioavailability of polychlorinated biphenyls (PCBs) from contaminated marine sediments. The sediments used were collected from 2 Superfund sites, New Bedford Harbor (MA, USA) and Gould Island (RI, USA). No sieving was conducted for either sediment after arrival, and sediments were stored in stainless steel drums at 2.8 to 4.0 °C in the dark until use. Exhaustive chemical extractions, single-point 24-h Tenax extractions, and 14-d bioaccumulation tests using the amphipod Leptocheirus plumulosus were conducted for both sediments. The sum of 119 PCB congeners from total exhaustive chemical extraction in the New Bedford Harbor and Gould Island sediments were 1084 and 188.2 µg/g organic carbon, respectively. The PCB concentrations from the bioaccumulation tests and Tenax extractions showed that both exposure metrics followed a similar trend in amount and distribution of PCB congeners. The results from both exposure metrics were fit into a log-log linear regression, and then compared with a previously developed log-log linear model for freshwater organisms. The results showed that although the marine data fell within the prediction intervals of the freshwater linear model, the marine regression followed a lower trajectory due to the differences in both the slopes and intercepts between the marine and freshwater regressions. The present study showed a strong relationship between Tenax and marine invertebrate PCB concentrations. Environ Toxicol Chem 2019;38:1188-1197. © 2019 SETAC.