Merck
  • UTX Affects Neural Stem Cell Proliferation and Differentiation through PTEN Signaling.
UTX Affects Neural Stem Cell Proliferation and Differentiation through PTEN Signaling.
Stem cell reports (2018-03-20)
Xuepei Lei, Jianwei Jiao
ABSTRACT
Neural stem cell (NSC) proliferation and differentiation in the developing brain is a complex process precisely regulated by intrinsic and extrinsic signals. Although epigenetic modification has been reportedly involved in the regulation of the cerebral cortex, whether UTX, an H3K27me3 demethylase, regulates the development of cerebral cortex during the embryonic period is unclear. In this study, we demonstrate that Utx deficiency by knockdown and conditional knockout increases NSC proliferation and decreases terminal mitosis and neuronal differentiation. Furthermore, we find that impairment of cortical development caused by lack of Utx is less significant in males than in females. In addition, UTX demethylates H3K27me3 at the Pten promoter and promotes Pten expression. P-AKT and P-mTOR levels are increased in the Utx conditional knockout cortices. Finally, Utx or Pten overexpression can rescue the impairment of brain development caused by Utx loss. These findings may provide important clues toward deciphering brain diseases.
Sigma-Aldrich
Anti-β-Tubulin III antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-UTX Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-PAX6 Antibody, from rabbit, purified by affinity chromatography