Merck
  • GNA13 expression promotes drug resistance and tumor-initiating phenotypes in squamous cell cancers.
GNA13 expression promotes drug resistance and tumor-initiating phenotypes in squamous cell cancers.
Oncogene (2017-12-20)
Suhail Ahmed Kabeer Rasheed, Hui Sun Leong, Manikandan Lakshmanan, Anandhkumar Raju, Dhivya Dadlani, Fui-Teen Chong, Nicholas B Shannon, Ravisankar Rajarethinam, Thakshayeni Skanthakumar, Ern Yu Tan, Jacqueline Siok Gek Hwang, Kok Hing Lim, Daniel Shao-Weng Tan, Paolo Ceppi, Mei Wang, Vinay Tergaonkar, Patrick J Casey, N Gopalakrishna Iyer
ABSTRACT
Treatment failure in solid tumors occurs due to the survival of specific subpopulations of cells that possess tumor-initiating (TIC) phenotypes. Studies have implicated G protein-coupled-receptors (GPCRs) in cancer progression and the acquisition of TIC phenotypes. Many of the implicated GPCRs signal through the G protein GNA13. In this study, we demonstrate that GNA13 is upregulated in many solid tumors and impacts survival and metastases in patients. GNA13 levels modulate drug resistance and TIC-like phenotypes in patient-derived head and neck squamous cell carcinoma (HNSCC) cells in vitro and in vivo. Blockade of GNA13 expression, or of select downstream pathways, using small-molecule inhibitors abrogates GNA13-induced TIC phenotypes, rendering cells vulnerable to standard-of-care cytotoxic therapies. Taken together, these data indicate that GNA13 expression is a potential prognostic biomarker for tumor progression, and that interfering with GNA13-induced signaling provides a novel strategy to block TICs and drug resistance in HNSCCs.
Sigma-Aldrich
Anti-GNA13 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GNA13 Mouse mAb (6F6-B5), liquid, clone 6F6-B5, Calbiochem®