Merck
  • Strategies for the synthesis of novel indole alkaloid-based screening libraries for drug discovery.
Strategies for the synthesis of novel indole alkaloid-based screening libraries for drug discovery.
Molecular diversity (2005-03-26)
Demosthenes Fokas, Libing Yu, Carmen M Baldino
ABSTRACT
A series of diverse indole-based chemotypes were synthesized from beta-tetrahydrocarboline (beta-THC) scaffolds prepared from commercially and readily available tryptamines and alpha-ketoesters. Diversity can be generated within these chemotypes through the following strategies: (a) appendage of substituents to the beta-THC scaffold, prepared in situ or as a template, through further elaboration and (b) skeletal modifications to the beta-THC scaffold via ring forming or ring breaking reactions. The strategies described here are amenable to high throughput solution-phase parallel synthesis, providing access to novel indole-based screening libraries for drug discovery.
Sigma-Aldrich
6-Fluorotryptamine hydrochloride, 99%