Merck
  • Hyaluronan-Based Nanohydrogels for Targeting Intracellular S. Aureus in Human Keratinocytes.
Hyaluronan-Based Nanohydrogels for Targeting Intracellular S. Aureus in Human Keratinocytes.
Advanced healthcare materials (2018-04-27)
Elita Montanari, Angela Oates, Chiara Di Meo, Josephine Meade, Rugiada Cerrone, Antonio Francioso, Deirdre Devine, Tommasina Coviello, Patrizia Mancini, Luciana Mosca, Pietro Matricardi
ABSTRACT
Staphylococcus aureus is one of the most significant human pathogens that is frequently isolated in a wide range of superficial and systemic infections. The ability of S. aureus to invade and survive within host cells such as keratinocytes and host immune cells has been increasingly recognized as a potential factor in persistent infections and treatment failures. The incorporation of antibiotics into hyaluronan-cholesterol nanohydrogels represents a novel paradigm in the delivery of therapeutic agents against intracellular bacteria. The work presented herein shows that NHs quickly enter human keratinocytes and accumulate into lysosomes. When used for targeting intracellular S. aureus the antimicrobial activity of loaded levofloxacin is enhanced, possibly changing the antibiotic intracellular fate from cytosol to lysosome. Indeed, gentamicin, an antibiotic that predominantly accumulates in lysosomes, shows significant and equal antibacterial activity when entrapped into NHs. These results strongly suggest that lysosomal formulations may display preferential activity toward intracellular S. aureus, opening new avenues for the use of HA-based NHs for treatment of such skin infections.
Sigma-Aldrich
4-Bromobutyric acid, 98%
Sigma-Aldrich
Triton X-100, laboratory grade