Merck
  • Matrix Metalloproteinase-9 and Tissue Inhibitor of Matrix Metalloproteinase-1 in Sepsis after Major Abdominal Surgery.
Matrix Metalloproteinase-9 and Tissue Inhibitor of Matrix Metalloproteinase-1 in Sepsis after Major Abdominal Surgery.
Disease markers (2018-06-05)
Suzana Bojic, Jelena Kotur-Stevuljevic, Aleksandra Aleksic, Jasna Gacic, Lidija Memon, Sanja Simic-Ogrizovic
ABSTRACT
The role of matrix metalloproteinase-9 (MMP-9) and tissue inhibitor of matrix metalloproteinase-1 (TIMP-1) in sepsis after major abdominal surgery and sepsis-associated organ dysfunction is unexplored. Fifty-three patients with sepsis after major abdominal surgery were compared to 50 operated and 50 nonoperated controls. MMP-9, TIMP-1, biomarkers of inflammation, kidney and liver injury, coagulation, and metabolic disorders were measured daily during 96 h following diagnosis of sepsis and once in controls. MMP-9/TIMP-1 ratios and disease severity scores were calculated. Use of vasopressors/inotropes, mechanical ventilation, and survival were recorded. Septic patients had lower MMP-9 and MMP-9/TIMP-1 ratios but higher TIMP-1 levels compared to controls. AUC-ROC for diagnosis of sepsis was 0.940 and 0.854 for TIMP-1 and 0.924 and 0.788 for MMP-9/TIMP-1 ratio (sepsis versus nonoperated and sepsis versus operated controls, resp.). Lower MMP-9 and MMP-9/TIMP-1 ratio and higher TIMP-1 levels were associated with shorter survival. MMP-9, TIMP-1, and MMP-9/TIMP-1 ratio correlated with biomarkers of inflammation, kidney and liver injury, coagulation, metabolic disorders, and disease severity scores. Use of vasopressors/inotropes was associated with higher TIMP-1 levels. MMP-9, TIMP-1, and MMP-9/TIMP ratio were good diagnostic or prognostic biomarkers of sepsis after major abdominal surgery and were linked to sepsis-associated organ dysfunction.
Sigma-Aldrich
Human HAVCR1 / Hepatitis A Virus Cellular Receptor 1 ELISA Kit