
Z-Aib-OH

Synonym(s):
Z-2-Methylalanine
Linear Formula:
C6H5CH2O2CNHC(CH3)2CO2H
CAS Number:
Molecular Weight:
237.25
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing