
Uzarin

Synonym(s):
Uzarigenin 3-sophoroside
Empirical Formula (Hill Notation):
C35H54O14
CAS Number:
Molecular Weight:
698.79
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing