
Triisobutylaluminum solution

Synonym(s):
TIBA solution
Linear Formula:
[(CH3)2CHCH2]3Al
CAS Number:
Molecular Weight:
198.32
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing