Skip to Content
Merck
Tocainide hydrochloride ≥98% (HPLC), solid

Tocainide hydrochloride

Tocainide hydrochloride ≥98% (HPLC), solid
Synonym(s):
2-Amino-N-(2,6-dimethylphenyl)propanamide hydrochloride
Empirical Formula (Hill Notation):
C11H16N2O · HCl
CAS Number:
Molecular Weight:
228.72
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing