Tetrahydropalmatine
Synonym(s):
2,3,9,10-Tetramethoxyberbine, 5,8,13,13a-Tetrahydro-2,3,9,10-tetramethoxy-6H-dibenzo[a,g]quinolizine, Berbine, THP
Empirical Formula (Hill Notation):
C21H25NO4
CAS Number:
Molecular Weight:
355.43
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing