![Tefluthrin certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/276/998/1a77f7a3-40ed-4608-b28c-dde06358bfb9/640/1a77f7a3-40ed-4608-b28c-dde06358bfb9.png)
Tefluthrin
![Tefluthrin certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/276/998/1a77f7a3-40ed-4608-b28c-dde06358bfb9/640/1a77f7a3-40ed-4608-b28c-dde06358bfb9.png)
Empirical Formula (Hill Notation):
C17H14ClF7O2
CAS Number:
Molecular Weight:
418.73
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2