
Spirotetramat Metabolite BYI08330 enol-glucoside

Synonym(s):
cis-3-(2,5-Dimethylphenyl)-8-methoxy-2-oxo-1-azaspiro [4.5]dec-3-en-4-yl β-D-glucopyranoside
Empirical Formula (Hill Notation):
C24H33NO8
CAS Number:
Molecular Weight:
463.52
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique