![Solifenacin for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/296/718/cca69a24-f7b2-4c72-9632-b7721c4dbf42/640/cca69a24-f7b2-4c72-9632-b7721c4dbf42.png)
Solifenacin for system suitability
![Solifenacin for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/296/718/cca69a24-f7b2-4c72-9632-b7721c4dbf42/640/cca69a24-f7b2-4c72-9632-b7721c4dbf42.png)
Synonym(s):
Solifenacin succinate
Empirical Formula (Hill Notation):
C23H26N2O2 · C4H6O4
CAS Number:
Molecular Weight:
480.55
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing