Skip to Content
Merck
Sodium D-3-hydroxybutyrate-2,4-13C2 endotoxin tested, ≥99 atom % 13C

Sodium D-3-hydroxybutyrate-2,4-13C2

Sodium D-3-hydroxybutyrate-2,4-13C2 endotoxin tested, ≥99 atom % 13C
Synonym(s):
(R)-(-)-3-Hydroxybutyric acid-2,4-13C2 sodium salt, Sodium (R)-3-hydroxybutyrate-2.4.13C2
Empirical Formula (Hill Notation):
13C2C2H7O3Na
Molecular Weight:
128.07

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Optical Isomer
Technique