![Sodium cromoglicate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/702/772/03f44567-b9ba-46ae-a8e7-4b19f62a870b/640/03f44567-b9ba-46ae-a8e7-4b19f62a870b.png)
Sodium cromoglicate
![Sodium cromoglicate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/702/772/03f44567-b9ba-46ae-a8e7-4b19f62a870b/640/03f44567-b9ba-46ae-a8e7-4b19f62a870b.png)
Synonym(s):
Cromolyn sodium salt, Cromoglycate, Cromoglycic acid
Linear Formula:
C23H14O11Na2
CAS Number:
Molecular Weight:
512.33
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing