Scopolamine N-oxide hydrobromide
Synonym(s):
(−)Scopolamine N-oxide hydrobromide, Hyoscine N-oxide hydrobromide
Empirical Formula (Hill Notation):
C17H21NO5 · HBr
CAS Number:
Molecular Weight:
400.26
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing