Picroside II
Synonym(s):
(1aS,1bS,2S,5aR,6S,6aS)-1a,1b,2,5a,6,6a-Hexahydro-6-[(4-hydroxy-3-methoxybenzoyl)oxy]-1a-(hydroxymethyl)oxireno[4,5]cyclopenta[1,2-c]pyran-2-yl-β-D-glucopyranoside, 6-Vanillylcatalpol, Amphicoside II, Vanilloyl catalpol
Empirical Formula (Hill Notation):
C23H28O13
CAS Number:
Molecular Weight:
512.46
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing