Phytanic acid
Synonym(s):
3,7,11,15-Tetramethylhexadecanoic acid, NSC 108698, Phytanoic acid
Empirical Formula (Hill Notation):
C20H40O2
CAS Number:
Molecular Weight:
312.53
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing