![Peimine phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/310/795/8a4067ec-958e-4cf9-b53e-4c0d4fcf886d/640/8a4067ec-958e-4cf9-b53e-4c0d4fcf886d.png)
Peimine
![Peimine phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/310/795/8a4067ec-958e-4cf9-b53e-4c0d4fcf886d/640/8a4067ec-958e-4cf9-b53e-4c0d4fcf886d.png)
Synonym(s):
(3β,5α,6α)-Cevane-3,6,20-triol, Dihydroisoimperialine, Verticine
Empirical Formula (Hill Notation):
C27H45NO3
CAS Number:
Molecular Weight:
431.65
Products
Formula Weight
Manufacturer
Product Line
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing