Skip to Content
Merck
Olsalazine sodium

Olsalazine sodium

Olsalazine sodium
Synonym(s):
Olsalazine sodium, 4,4′-Dihydroxyazobenzene-3,3′-dicarboxylic acid disodium salt, 5,5′-Azobis(salicylic acid, sodium salt), Mordant Yellow 5
Empirical Formula (Hill Notation):
C14H8N2Na2O6
CAS Number:
Molecular Weight:
346.20

Products

Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing