
N-p-Chlorobenzoyltyramine

Synonym(s):
4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide
Empirical Formula (Hill Notation):
C15H14ClNO2
CAS Number:
Molecular Weight:
275.73
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing