N-Nitroso-bisoprolol
Synonym(s):
1-{4-{[2-(1-Methylethoxy)ethoxy]methyl}phenoxy}-3-[(1-methylethyl)nitrosoamino]-2-propanol
CAS Number:
Products
Brand
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing