Skip to Content
Merck
N-Lactoyl ethanolamine ≥97%, FG

N-Lactoyl ethanolamine

N-Lactoyl ethanolamine ≥97%, FG
Synonym(s):
N-Hydroxyethyllactamide
Empirical Formula (Hill Notation):
C5H11NO3
CAS Number:
Molecular Weight:
133.15

Products

Product Category
Brand
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing