![N-Acetyl-2,3-dehydro-2-deoxyneuraminic acid ≥93% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/384/988/dd168ded-944d-4b64-a9da-06eb62cd467f/640/dd168ded-944d-4b64-a9da-06eb62cd467f.png)
N-Acetyl-2,3-dehydro-2-deoxyneuraminic acid
![N-Acetyl-2,3-dehydro-2-deoxyneuraminic acid ≥93% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/384/988/dd168ded-944d-4b64-a9da-06eb62cd467f/640/dd168ded-944d-4b64-a9da-06eb62cd467f.png)
Synonym(s):
2,3-Dehydro-2-deoxy-N-acetylneuraminic acid
Empirical Formula (Hill Notation):
C11H17NO8
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing