Molybdenum oxide concentrate
Synonym(s):
Molybdenum(VI) oxide, Molybdenum trioxide
Linear Formula:
MoO3
CAS Number:
Molecular Weight:
143.94
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing