![Mirtazapine for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/286/305/bee5bf42-6684-4688-8721-2ff73700c5e2/640/bee5bf42-6684-4688-8721-2ff73700c5e2.png)
Mirtazapine for system suitability
![Mirtazapine for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/286/305/bee5bf42-6684-4688-8721-2ff73700c5e2/640/bee5bf42-6684-4688-8721-2ff73700c5e2.png)
Synonym(s):
Mirtazapine, 1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine
Empirical Formula (Hill Notation):
C17H19N3
CAS Number:
Molecular Weight:
265.35
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing