
Levomepromazine maleate

Empirical Formula (Hill Notation):
C23H28N2O5S
CAS Number:
Molecular Weight:
444.54
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing